Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R882A-05N

Sigma-Aldrich

Rat Astrocytes: RA, neonatal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

rat brain (Rat brain)

Quality Level

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

morphology

glial

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

seizures

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.


Rat astrocytes (RA) are derived from rat cerebral cortex. Astrocytes are the most abundant cells in the central nervous system where they perform many functions, such as providing mechanical support and nutrients to neurons and removal of wastes from neurons; providing signaling to endothelial cells; regulating neurogenesis and controlling synaptic function. As the recognition of the importance of astrocytes in nervous system is increasing, RA serve as a useful in vitro model for exploring the diversity of astrocytes functions.Characterization: Stained positive for GFAP.Rat Astrocytes (RA) and Rat Brain Microvascular Endothelial Cells (RBMEC), both from Cell Applications, Inc., have been used to assess neuroprotective capabilities of bioenergy stabilizers in an in vitro model of stroke (Liu, 2013) and to investigates the effects of astrocytes on brain microvascular endothelial cells in relation to iron transport pathway (McCarthy, 2014).

Cell Line Origin

Brain

Application

Neuroscience research, mechanical & biochemical support, ion balance, nutrient provision, waste removal, cell signaling, regulation of neurogenesis, control of synaptic function, astrocyte function, gene expressino, injury repair, gliotransmitter release, neuronal excitability.

Components

Basal Medium containing 10% FBS & 10% DMSO

Quality

Each lot was tested for proper morphology, Population Doublings, Negative for HIV, Hepatitis B, Hepatitis C, mycoplasma, bacteria, and fungi.

Preparation Note

  • 2nd passage, >500,000 cells in Basal Medium containing 10% FBS & 10% DMSO
  • Can be cultured at least 10 doublings

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service