Key Documents
R0401
NovaRack
red
About This Item
UNSPSC Code:
41122804
NACRES:
NB.35
packaging
pack of 1 ea
manufacturer/tradename
Diversified Biotech NORC-1000
color
red
General description
NovaRack provides universal housing for test tubes and vials of different sizes on the same surface. The numeric labeling for each tube cavity allows one to easily organize and track up to 46 tubes. It accommodates 24 microcentrifuge tubes (0.5-2 mL), 14 test tubes (12 x 75 mm culture tubes, cryovials and a variety of blood collection tubes), 7 centrifuge tubes (15 mL) and one 50 mL tube. NovaRack is constructed of polypropylene, is autoclavable and is available in five colors. With the addition of the Carousel Extension rod, the space saving ability is doubled and the top rack is allowed to spin.
Legal Information
Novarack is a trademark of Diversified Biotech
Choose from one of the most recent versions:
Certificates of Analysis (COA)