Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P9107

Sigma-Aldrich

Anti-Potassium Channel Kv1.3 (Charybdotoxin-Sensitive Voltage-Dependent Potassium Channel, HLK3) antibody produced in rabbit

IgG fraction of antiserum, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat, mouse, human

technique(s)

immunocytochemistry: 1:50 using cells permeabilized with 0.2% Triton X-100
western blot (chemiluminescent): 1:100

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNA3(3738)
mouse ... Kcna3(16491)
rat ... Kcna3(29731)

Specificity

Recognizes a full-length Kv1.3 protein. Does not cross-react with other potassium channel antigens.

Immunogen

highly purified GST fusion protein from the C-terminal of human potassium channel Kv1.3. The protein corresponds to amino acids 471-523.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Physical form

Lyophilized at ~1 mg/ml from phosphate buffered saline, pH 7.4, containing 5% sucrose, 1% bovine serum albumin, and 0.025% sodium azide.

Preparation Note

Protein A purified.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stella A Nicolaou et al.
Journal of immunology (Baltimore, Md. : 1950), 183(10), 6296-6302 (2009-10-21)
The immunological synapse (IS), a highly organized structure that forms at the point of contact between a T cell and an APC, is essential for the proper development of signaling events, including the Ca(2+) response. Kv1.3 channels control Ca(2+) homeostasis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service