Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P8366

Sigma-Aldrich

Nunclon® MicroWell plates for automation

96 well (with lid), flat bottom clear wells

Synonym(s):

automation 96 well plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.22
material:
clear polystyrene
flat bottom clear wells
polystyrene
size:
96 wells
sterility:
sterile
binding type:
Nunclon Delta surface
Tissue Culture (TC)-treated surface
feature:
lid

material

clear polystyrene
flat bottom clear wells
polystyrene

sterility

sterile

feature

lid

packaging

pack of 50 (individually wrapped)

manufacturer/tradename

Nunc 167008

maximum volume

400 μL

size

96 wells

well maximum volume

250 μm

working volume

250 μL

binding type

Nunclon Delta surface
Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Standard 96 well footprint meets the SBS guidelines
  • High Flange ideal for robotics and automation
  • Nunclon Delta surface ensures consistent optimal cell attachment and growth
  • Features a tight closing lid that ensures sample protection and prevents evaporation
  • Raised rim minimizes cross-contamination risk
  • Stackable - A convenient space-saving option

Legal Information

Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service