Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

P1253

Sigma-Aldrich

Phosvitin from egg yolk

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.81

biological source

egg yolk

form

powder

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Phosvitin from egg yolk is a polyanionic phosphoserine rich protein. This phosphoglycoprotein comprises α-helices and β-sheets structural elements, which are poorly organized due to the presence of phosphoserine. It represents about 11% of yolk proteins and is hydrophilic.

Application

Phosvitin from egg yolk has been used:
  • as a control protein for immobilization on-chip for deep purple protein staining methods for monitoring post-translation modifications
  • as a substrate for casein kinase Iε in in vitro casein kinase assay
  • as a standard in gel filtration chromatography (GFC) and sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE) for quantification of phosvitin from egg extracts
  • for immobilization onto Layer-by-Layer (LbL) for Fourier transform infrared spectroscopy-attenuated total reflectance (FTIR-ATR) studies

Biochem/physiol Actions

Phosvitin fed to rats by adding egg yolk to their food resulted in decreased absorption of magnesium, iron and calcium but not of phosphorus.
Phosvitin from egg yolk is a calcium chelator and binds to iron present in the yolk. The metal-binding functionality is exploited in mineral-binding bioactive peptide production. It is a nutraceutical and has potential for phosphopeptides production.

Other Notes

A phosophoprotein containing 8-10% phosphorus. Molar N/P ratio approx. 2.7.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Himali Samaraweera et al.
Journal of food science, 76(7), R143-R150 (2011-08-03)
Phosphopeptides are among the most interesting biomolecules with characteristic molecular structure and functions. They usually contain clusters of phosphoserines, which can effectively bind calcium and iron, and inhibit formation of insoluble calcium phosphates or iron complexes. Therefore, phosphopeptides can increase
Samooel Jung et al.
Food chemistry, 135(3), 993-998 (2012-09-08)
Phosvitin is a phosphoglycoprotein present in egg yolk. More than half of the amino acids in phosvitin molecule are serine, of which >90% are phosphorylated. Therefore, phosvitin has a strong metal binding capability. The aim of this study was to
Shuichi Ito et al.
Journal of biomedical materials research. Part A, 100(10), 2760-2765 (2012-05-25)
The aim of this study is to evaluate the mineralizing potential of acidic monomers and their calcium salts for mineralization, using an in vitro mineral induction model. Phosvitin (PV) was used as a model phosphoprotein in this study. PV was
K Y Ko et al.
Poultry science, 90(5), 1096-1104 (2011-04-15)
The objective of this study was to develop a new protocol that could be used for large-scale separation of phosvitin from egg yolk using ethanol and salts. Yolk granules, which contain phosvitin, were precipitated after diluting egg yolk with 9
S-I Ishikawa et al.
Journal of food science, 72(6), S412-S419 (2007-11-13)
Egg yolk decreases the absorption of iron. The effects of egg yolk protein and egg yolk phosvitin on the absorption of calcium, magnesium, and iron were investigated by in vivo studies. Male Wistar rats were fed purified diets containing casein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service