Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N8164

Sigma-Aldrich

NSC727447

≥98% (HPLC)

Synonym(s):

2-Amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylic acid amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2OS
CAS Number:
Molecular Weight:
210.30
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: >10 mg/mL

storage temp.

room temp

SMILES string

NC(=O)c1c(N)sc2CCCCCc12

InChI

1S/C10H14N2OS/c11-9(13)8-6-4-2-1-3-5-7(6)14-10(8)12/h1-5,12H2,(H2,11,13)

InChI key

LULYZYNTDPUEKK-UHFFFAOYSA-N

Biochem/physiol Actions

NSC727447 is an inhibitor of ribonuclease H (Rnase H) of HIV-1 and HIV-2. NSC727447 has little activity against E. coli RNase H, but great selectivity over human Rnase H.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service