Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N6163

Sigma-Aldrich

Anti-Nerve Growth Factor Receptor/TNFRSF16 antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-LNGFR, Anti-Ngfr, Anti-Tnfrsf16, Anti-p75

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... NGFR(4804)

Looking for similar products? Visit Product Comparison Guide

General description

Neurotrophins (NT) are growth factors that are essential for the development and maintenance of the nervous system. One of the receptors that mediate the effects of NTs is the Nerve Growth Factor Receptor/TNFRSF16 (p75NGFR, p75NTR). Nerve growth factor (NGF) binds to either to NGF receptor (NGFR) or the Trk receptor and can elicit biological responses through each of the receptors independently. NGFR mediates the effects of NGF that are important in the survival and differentiation of neurons and neural crest-derived cells during embryonic and early postnatal development and maintenance of peripheral neurons. Signaling of NGF through NGFR is also implicated in inflammation and allergic reactions. Expression of NGFR has been associated with favourable prognosis and survival in breast and pancreatic cancers
Anti-Nerve Growth Factor Receptor/TNFRSF16 recognizes human NGF R.

Immunogen

purified, Sf21-derived recombinant human p75 neurotrophin R extracellular domain.

Application

Anti-Nerve Growth Factor Receptor/TNFRSF16 antibody may be used for immunoblotting at a working concentration of 0.1-0.2 μg/ml.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wolfgang Andreas Nockher et al.
The Journal of allergy and clinical immunology, 117(3), 583-589 (2006-03-09)
Understanding the complex pathophysiology of allergic diseases has been a main challenge of clinical and experimental research for many years. It is well known that the allergic inflammation triggers neuronal dysfunction and structural changes in the diseased tissues such as
Y Indo
Clinical genetics, 82(4), 341-350 (2012-08-14)
Congenital insensitivity to pain with anhidrosis (CIPA) is an autosomal recessive genetic disorder characterized by insensitivity to pain, anhidrosis (the inability to sweat) and mental retardation. Nerve growth factor (NGF) is a well-known neurotrophic factor essential for the survival and
Yong Zhang et al.
Oncology reports, 14(1), 161-171 (2005-06-10)
Nerve growth factor (NGF), the prototypic member of the neurotrophin family of growth factors, exerts both stimulatory and inhibitory effects on neuronal and certain non-neuronal tumors, depending on the type of tumor. It has been reported that two types of
C D Mills et al.
European journal of pain (London, England), 17(4), 469-479 (2012-08-24)
Injection of nerve growth factor (NGF) produces mechanical and thermal hypersensitivity in rodents and humans. Treatment with sequestering antibodies demonstrates the importance of NGF in various pain states, with efficacy seen in a number of animal pain models and in
Hrishikesh M Mehta et al.
Journal of neuroscience research, 90(12), 2259-2271 (2012-08-21)
Nerve growth factor (NGF) is a homodimer that binds to two distinct receptor types, TrkA and p75, to support survival and differentiation of neurons. The high-affinity binding on the cell surface is believed to involve a heteroreceptor complex, but its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service