Skip to Content
MilliporeSigma
All Photos(1)

Documents

N1895

Sigma-Aldrich

4-Nitrophenyl α-D-xylopyranoside

α-xylosidase substrate, ≥99% (HPLC), powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13NO7
CAS Number:
Molecular Weight:
271.22
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

product name

4-Nitrophenyl α-D-xylopyranoside, α-xylosidase substrate

Assay

≥99% (HPLC)

form

powder

solubility

methanol: soluble 20 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

OC1COC(Oc2ccc(cc2)N(=O)=O)C(O)C1O

InChI

1S/C11H13NO7/c13-8-5-18-11(10(15)9(8)14)19-7-3-1-6(2-4-7)12(16)17/h1-4,8-11,13-15H,5H2

InChI key

MLJYKRYCCUGBBV-UHFFFAOYSA-N

Substrates

Chromogenic substrate for α-xylosidase

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Vitor Trinca et al.
iScience, 26(4), 106449-106449 (2023-04-07)
Soil biota has a crucial impact on soil ecology, global climate changes, and effective crop management and studying the diverse ecological roles of dipteran larvae deepens the understanding of soil food webs. A multi-omics study of Pseudolycoriella hygida comb. nov.
Annette Biederbick et al.
European journal of cell biology, 82(1), 19-29 (2003-02-27)
Serglycin is known as a secretory granule proteoglyean in hematopoietic cells. In this study we identified a high-molecular-weight molecule in aggregated content proteins of zymogen granules of pancreatic acinar cells. The amino acid composition of the isolated protein showed high
S G Venkatesh et al.
American journal of physiology. Cell physiology, 283(2), C438-C445 (2002-07-11)
Sulfated proteoglycans have been proposed to play a role in the sorting and storage of secretory proteins in exocrine secretory granules. Rat parotid acinar cells expressed a 40- to 60-kDa proteoglycan that was stored in secretory granules. Treatment of the
Talia Weinstein et al.
Connective tissue research, 53(2), 169-179 (2011-12-14)
The familial disease of hereditary multiple exostoses is characterized by abnormal skeletal deformities requiring extensive surgical procedures. In hereditary multiple exostoses patients there is a shortage in the pericellular glycosaminoglycan (GAG) of heparan sulfate (HS), related to defective activity of
M Gómez et al.
Biochimie, 83(10), 961-967 (2001-12-01)
The variation of kinetic parameters of beta-xylosidase from Trichoderma reesei QM 9414 with pH was used to elucidate the chemical mechanism of the p-nitrophenyl beta-D-xylopyranoside hydrolysis. The pH-dependence of V and V/K(m) showed that a group on the enzyme with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service