Skip to Content
MilliporeSigma

Skip To

MTOX1024

HepaRG Cells PERK (-/-)

human female liver (hepatocarcinoma and hepatitis C tumor)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 vial
Usually ships in 3 weeks.
$1,040.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.81
Biological source:
human female liver (hepatocarcinoma and hepatitis C tumor)

$1,040.00


Usually ships in 3 weeks.

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

HepaRG Cells PERK (-/-), one vial

biological source

human female liver (hepatocarcinoma and hepatitis C tumor)

Quality Segment

form

liquid

OMIM accession no.

storage temp.

−196°C

Gene Information

human ... EIF2AK3(9451)

General description

HepaRG is a human hepatoma cell line. The cells possess a pseudodiploid karyotype and have been characterized as an oval ductular bipotent hepatic cell line as they have the ability to differentiate into both biliary and hepatocyte lineages in the presence of DMSO. HepaRG PERK knockout cells express the major xenobiotic sensors (PXR, CAR and AhR), drug transporters, phase I and II drug metabolizing enzymes as well as key hepatic transcription factors involved in stress response pathways.

Application

See technical bulletin for detailed protocols

Features and Benefits

HepaRG cells are the most metabolically active human hepatocyte cell line developed to date. These the cells are suitable for a wide variety of studies for drug metabolism, CYP induction, metabolism-mediated toxicity, transporter, and hepatotoxicity. - Sigma′s HepaRG PERK Knockout (KO) allows investigations of Wolcott-Rallison syndrome and endoplasmic reticulum stress response involving PERK in the liver.

Analysis Note

Tested for Mycoplasma, sterility, post-freeze viability, short terminal repeat (STR) analysis for cell line identification, cytochrome oxidase I (COI) analysis for cell line species confirmation.

Legal Information

Exhibit 1: ADME/Tox cell lines license
Exhibit 2: HepaRG limited use license
HepaRG is a trademark of BioPredic International company

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MTOX1011MTOX1020MTOX1013
biological source

human female liver (hepatocarcinoma and hepatitis C tumor)

biological source

human female liver (Source Disease: Hepatocarcinoma and Hepatitis C)

biological source

human female liver (hepatocarcinoma and hepatitis C tumor)

biological source

human female liver (hepatocarcinoma and hepatitis C tumor)

storage temp.

−196°C

storage temp.

−196°C

storage temp.

−196°C

storage temp.

−196°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

OMIM accession no.

604032

OMIM accession no.

603065

OMIM accession no.

601107

OMIM accession no.

Gene Information

human ... EIF2AK3(9451)

Gene Information

human ... NR1I2(1337040)

Gene Information

human ... ABCC2(1244)

Gene Information

human ... AHR(196)

form

liquid

form

liquid

form

liquid

form

liquid


Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters