Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAS008

Sigma-Aldrich

Urea (BUN) Quick Test Strips

Sufficient for 10 tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106
NACRES:
NA.84

input

urine
serum
beverage(s)
food(s)
plasma

application(s)

food and beverages

detection method

colorimetric

storage temp.

2-8°C

General description

Urea, the primary end product of protein breakdown in animals, is mainly synthesized in the liver and excreted by the kidneys. It plays a crucial role in eliminating toxic ammonia from the body. Widely utilized in agriculture as a fertilizer and in animal feed, urea levels in milk are monitored to assess dietary protein intake for animals. Additionally, trace amounts of urea are present in various fermented foods and beverages. Assessing urea levels is valuable in biochemical and biomedical research, particularly for evaluating kidney function. Elevated urea levels are commonly linked to conditions like nephritis, renal ischemia, urinary tract obstruction, and certain extrarenal ailments such as congestive heart failure, liver diseases, and diabetes. Conversely, decreased levels may indicate acute hepatic insufficiency, although they can also result from overly aggressive parenteral fluid therapy.

Application

The Urea (BUN) Quick Test Strips are designed for determining urea levels in the following applications:
  • Food and Beverage Testing
  • Agriculture Testing
  • Biological samples such as plasma, serum, and urine.

Features and Benefits

  • Rapid and Sensitive: Provides swift results using just 20 μL of sample for semi-quantitative measurement across a range of 0-1500 mg/L (undiluted) Urea, ensuring quick and accurate analysis.
  • Efficient: Sample treatment and assay completion take less than 15 minutes, saving time and enhancing productivity.
  • User-Friendly: Convenient test strip design removes the need for a plate or cuvette reader, simplifying the testing process and making it accessible for users of all skill levels.

Suitability

This kit is suitable for the detection Urea (BUN) in a variety of food and beverage samples.

Principle

The Urea (BUN) Quick Test Strips are based on the urease catalyzed conversion of urea to ammonium and carbon dioxide. The pH of the reaction medium is monitored by a chromogen and the intensity of the product color is directly proportional to the urea concentration in the sample. The Urea (BUN) Quick Test strips allow for rapid and inexpensive quantitative determination of urea or blood urea nitrogen (BUN) and does not require sophisticated laboratory instruments.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service