Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK446

Sigma-Aldrich

Acid Phosphatase Assay Kit

Sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

Quality Level

input

serum
plasma
tissue sample(s)
cell lysate

detection method

colorimetric

relevant disease(s)

cancer, prostate tumor; gastrointestinal diseases

storage temp.

−20°C

General description

Acid Phosphatase (ACP) is an enzyme which catalyzes the cleavage of phosphate groups from other molecules during digestion. Acid phosphatase can be found in lysosomes and become active after fusing with endosomes, acidifying the pH and, thus, creating an optimal environment for ACP. ACP can also be found in bone, spleen, liver, kidney and blood. Serum levels can be used as a biomarker for prostatic carcinoma, although prostate-specific antigen (PSA) is more widely used.

Application

The Acid Phosphatase Assay Kit may be used for:
  • Cancer Research
  • Gastrointestinal Research
  • Metabolism Research

Features and Benefits

Efficient and Precise: Requires only 20 μL of sample with a 30-minute assay reaction to detect acid phosphatase levels across a broad range from 0.05 to 60 U/L, providing both convenience and sensitivity.

Compatibility with High-Throughput Systems: Easily incorporate our kit into high-throughput handling systems, ensuring smooth and accurate processing, enhancing efficiency in your laboratory workflow.

Suitability

The kit is suitable for the determination of acid phosphatase in biological samples (e.g. plasma, serum, cell lysate, tissue samples.)

Principle

The non-radioactive, colorimetric Acid Phosphatase Assay Kit is based on the cleavage of p-nitrophenol from the synthetic substrate. p-Nitrophenol becomes intensely colored after addition of the stop reagent. The increase in absorbance at A405 nm after addition of the stop reagent is directly proportional to the enzyme activity.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service