Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK438

Sigma-Aldrich

Glutamine Assay Kit

Sufficient for 100 colorimetric tests

Synonym(s):

Glutamine Quantification Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

Quality Level

application(s)

pharmaceutical

detection method

colorimetric

relevant disease(s)

immunological diseases; cancer; gastrointestinal diseases

storage temp.

−20°C

General description

Glutamine is an amino acid synthesized in the muscle that plays major roles in protein synthesis, acid-base balance, anabolic processes and is utilized for cellular energy and as a carbon source. It is used in the treatment of injury, trauma, burns, and also as a supplement for muscle growth and post-surgery healing.

Application

The Glutamine Assay Kit may be used for:
  • Cancer Research
  • Immunological Research
  • Gastrointestinal Disease Research

Features and Benefits

Broad Detection Range: Detect Glutamine levels accurately across a wide range, from 0.023 to 2 mM, using only 20 μL of sample in a 96-well plate setup, ensuring comprehensive analysis with minimal sample requirement.

Simplified Process: Experience a streamlined process with the addition of only a single working reagent and a room temperature reaction, reducing complexity and saving valuable time and effort.

Compatibility with High-Throughput Systems: Easily incorporate our kit into high-throughput handling systems, ensuring smooth and accurate processing, enhancing efficiency in your laboratory workflow.

Suitability

The kit is suitable for the quantitative determination of glutamine and evaluation of drug effects on glutamine metabolism in serum, plasma, urine, cells, tissue etc.

Principle

Simple, direct and automation-ready procedures for measuring glutamine concentration are very desirable. The Glutamine Assay Kit is based on the hydrolysis of glutamine to glutamate and the colorimetric determination of the product. The intensity of the product color, measured at A565 nm, is proportional to the glutamine concentration in the sample.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service