Skip to Content
MilliporeSigma
All Photos(1)

Documents

M9267

Supelco

Myoglobin from equine heart

Isoelectric focusing marker, pI (1) 6.8, (2) 7.2

Synonym(s):

Myoglobin from horse heart

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41105336
NACRES:
NA.25

Quality Level

form

powder

pI 

(1) 6.8, (2) 7.2

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Myoglobin from equine heart has been used as a pI marker in two-dimensional gel electrophoresis.

Biochem/physiol Actions

Myoglobin from equine heart is a protein which is used as a standard pI marker for isoelectric focussing experiments.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Purification and some properties of a protopectin-solubilizing enzyme that has potent activity on sugar beet protopectin
Takuo S and Sakamoto T
Agricultural and Biological Chemistry, 54, 879-889 (1990)
Two-dimensional gel isoelectric focusing.
Stastna M and Slais K
Electrophoresis, 26, 3586-3591 (2005)
Forskolin and camptothecin induce a 30 kDa protein associated with melatonin production in Y79 human retinoblastoma cells.
Janavs JL, et al.
The Journal of Neuroscience, 15, 298-309 (1995)
Yie-Hwa Chang et al.
Vaccines, 8(3) (2020-07-30)
Francisella tularensis (F. tularensis) is an intracellular pathogen that causes a potentially debilitating febrile illness known as tularemia. F. tularensis can be spread by aerosol transmission and cause fatal pneumonic tularemia. If untreated, mortality rates can be as high as
Ying Guo et al.
BMC microbiology, 8, 226-226 (2008-12-19)
Iron is recognized as an important trace element, essential for most organisms including pathogenic bacteria. HugZ, a protein related to heme iron utilization, is involved in bacterial acquisition of iron from the host. We previously observed that a hugZ homologue

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service