Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M7820

Sigma-Aldrich

Anti-MAGE-C1 antibody, Mouse monoclonal

clone CT7-33, purified from hybridoma cell culture

Synonym(s):

Anti-CT7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

CT7-33, monoclonal

form

buffered aqueous solution

mol wt

antigen ~180 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1-2 μg/mL using total cell extract of SKML37 cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAGEC1(9947)

General description

Anti-MAGE-C1 antibody, Mouse monoclonal (mouse IgG1 isotype) is derived from the hybridoma CT7-33 produced by the fusion of mouse myeloma cells (SP2/0 cells) and splenocytes from mice immunized with human recombinant MAGE-C1. The first melanoma-associated antigen (MAGE) to be identified was MAGE-A1. The MAGE family of proteins contains over 25 genes that have now been identified in human. These proteins share a homologous region, named the MAGE homology domain (MHD).
MAGE family member C1 (MAGE-C1) belongs to the melanoma antigen gene (MAGE) family and consists of 1142 amino acids. The gene encoding MAGE-C1 is localized on human chromosome Xq26-27 and has four exons.

Immunogen

human recombinant MAGE-C1.

Application

  • Anti-MAGE-C1 antibody, Mouse monoclonal has been used in:
  • enzyme-linked immunosorbent assay (ELISA)
  • immunocytochemistry
  • immunoblotting
  • immunoprecipitation
  • immunohistochemistry

Biochem/physiol Actions

Although the physiological role of melanoma-associated antigen (MAGE) proteins is unknown, it has been reported that MAGE-C1 interacts and co-localizes with an additional cancer testis antigen, NY-ESO-1, in melanoma cells.
MAGE family member C1 (MAGE-C1) has been studied as the most expressed cancer or testis antigen in multiple myeloma.

Target description

MAGE-C1 is a member of the melanoma antigen gene (MAGE) family. The proteins of this family are tumor-specific antigens that can be recognized by autologous cytolytic T lymphocytes.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kirsty Wienand et al.
PloS one, 10(3), e0120734-e0120734 (2015-03-21)
The malignant cell phenotype of Multiple Myeloma (MM) remains unclear with studies proposing it to be either clonotypic B or proliferating plasma cells. Cancer/testis antigen MAGE C1 is being extensively studied in MM and it has been suggested that it
MAGE-E1, a new member of the melanoma-associated antigen gene family and its expression in human glioma
Sasaki M, et al.
Cancer Research, 61(12), 4809-4814 (2001)
Physical interaction of two cancer-testis antigens, MAGE-C1 (CT7) and NY-ESO-1 (CT6)
Cho HJ, et al.
Cancer Immunity, 6(1), 12-12 (2006)
Achim A Jungbluth et al.
International journal of cancer, 99(6), 839-845 (2002-07-13)
CT7 (MAGE-C1) is a member of the cancer testis (CT) antigen family. The present study describes the generation of CT7-33, a monoclonal antibody (MAb) to CT7, and the preliminary protein expression analysis of CT7 in normal tissues and in a
Henna Mattila et al.
BMC cancer, 11, 327-327 (2011-08-04)
Several predisposition loci for hereditary prostate cancer (HPC) have been suggested, including HPCX1 at Xq27-q28, but due to the complex structure of the region, the susceptibility gene has not yet been identified. In this study, nonsense-mediated mRNA decay (NMD) inhibition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service