Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M1692

Sigma-Aldrich

MRS 1845

≥95% (HPLC), solid

Synonym(s):

N-Propargylnitrendipene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22N2O6
CAS Number:
Molecular Weight:
398.41
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥95% (HPLC)

form

solid

color

yellow

solubility

DMSO: soluble ≥10 mg/mL at 60 °C
H2O: insoluble

storage temp.

2-8°C

SMILES string

CCOC(=O)C1=C(C)N(CC#C)C(C)=C(C1c2cccc(c2)[N+]([O-])=O)C(=O)OC

InChI

1S/C21H22N2O6/c1-6-11-22-13(3)17(20(24)28-5)19(18(14(22)4)21(25)29-7-2)15-9-8-10-16(12-15)23(26)27/h1,8-10,12,19H,7,11H2,2-5H3

InChI key

BITHABUTZRAUGT-UHFFFAOYSA-N

Biochem/physiol Actions

Store-operated calcium (SOC) channel inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bo-Hong Lin et al.
Environmental toxicology, 31(11), 1293-1306 (2016-10-19)
Smoking increases the risk of cardiovascular disorders and leads to damage caused by inflammation and oxidative stress. The actin cytoskeleton is a key player in the response to inflammatory stimuli and is an early target of cellular oxidative stress. The
Rolando Berlinguer-Palmini et al.
PloS one, 8(11), e82180-e82180 (2013-12-07)
Limited information is available on the brain expression and role of GPR35, a Gi/o coupled receptor activated by kynurenic acid (KYNA). In mouse cultured astrocytes, we detected GPR35 transcript using RT-PCR and we found that KYNA (0.1 to 100 µM)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service