Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

L5793

Sigma-Aldrich

Leupeptin

powder, ≥95% (HPLC)

Synonym(s):

Leupeptin hemisulfate salt, Acetyl-Leu-Leu-Arg-al, N-Acetyl-L-leucyl-L-leucyl-L-argininal hemisulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C20H38N6O4 · 1/2H2SO4
CAS Number:
Molecular Weight:
475.59
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

product name

Leupeptin, BioUltra, microbial, ≥95% (HPLC)

biological source

microbial

Quality Level

product line

BioUltra

Assay

≥95% (HPLC)

form

powder

solubility

H2O: 50 mg/mL

storage temp.

−20°C

SMILES string

OS(O)(=O)=O.CC(C)C[C@H](NC(C)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCNC(N)=N)C=O.CC(C)C[C@H](NC(C)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCNC(N)=N)C=O

InChI

1S/2C20H38N6O4.H2O4S/c2*1-12(2)9-16(24-14(5)28)19(30)26-17(10-13(3)4)18(29)25-15(11-27)7-6-8-23-20(21)22;1-5(2,3)4/h2*11-13,15-17H,6-10H2,1-5H3,(H,24,28)(H,25,29)(H,26,30)(H4,21,22,23);(H2,1,2,3,4)/t2*15-,16-,17-;/m00./s1

InChI key

CIPMKIHUGVGQTG-VFFZMTJFSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibitor of serine and cysteine proteases. Inhibits plasmin, trypsin, papain, calpain, and cathepsin B. Does not inhibit pepsin, cathepsins A and D, thrombin, or α-chymotrypsin. Effective concentration 10-100 μM. There have been numerous studies using leupeptin to protect against hearing loss caused by acoustic overstimulation or the ototoxic antibiotic gentamicin. (Loss of cochlear hair cells is believed to be mediated by calpain.)

Analysis Note

Leupeptin gives multiple peaks on HPLC because of the formation of tautomeric isomers in solution. Majority of contaminating peptide is racemized leupeptin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Massimo Ganassi et al.
Molecular cell, 63(5), 796-810 (2016-08-30)
Stress granules (SGs) are ribonucleoprotein complexes induced by stress. They sequester mRNAs and disassemble when the stress subsides, allowing translation restoration. In amyotrophic lateral sclerosis (ALS), aberrant SGs cannot disassemble and therefore accumulate and are degraded by autophagy. However, the
Lifeng Fu et al.
mBio, 12(5), e0222021-e0222021 (2021-09-29)
Coronavirus disease 2019 (COVID-19) has caused huge deaths and economic losses worldwide in the current pandemic. The main protease (Mpro) of severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) is thought to be an ideal drug target for treating COVID-19. Leupeptin
Qinggong Wang et al.
Nature communications, 13(1), 454-454 (2022-01-26)
Melatonin receptors (MT1 and MT2 in humans) are family A G protein-coupled receptors that respond to the neurohormone melatonin to regulate circadian rhythm and sleep. Numerous efforts have been made to develop drugs targeting melatonin receptors for the treatment of
Zhiwei Chen et al.
Oncogenesis, 9(5), 49-49 (2020-05-15)
Carbidopa, a peripheral decarboxylase inhibitor used with L-DOPA to treat Parkinson's disease, has attracted significant interest in recent years for its anticancer effect. Increasing evidence reveals that Carbidopa can inhibit cancer cell growth and induce apoptosis through aryl hydrocarbon receptor
Rafal P Piprek et al.
Differentiation; research in biological diversity, 106, 23-34 (2019-03-11)
The extracellular matrix (ECM) proteins play an important role in the establishment of the sex-dependent structure of developing gonads. The matrix metalloproteinases (MMPs) are the major players in the regulation of ECM. Our hypothesis was that the MMPs-dependent regulation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service