Skip to Content
MilliporeSigma

Skip To

I1764

Insulin Chain B Oxidized from bovine pancreas

BioReagent, suitable for protein sequencing

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 vial

Available to ship TODAYfromMILWAUKEE

$70.20
$59.67

About This Item

Empirical Formula (Hill Notation):
C157H232N40O47S2
CAS Number:
Molecular Weight:
3495.89
UNSPSC Code:
12352202
NACRES:
NA.26
MDL number:

$59.67

List Price$70.20Save 15%
Web-Only Promotion

Available to ship TODAYDetails


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Segment

packaging

vial of 100 μg

technique(s)

protein sequencing: suitable

UniProt accession no.

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(O)C[C@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CNC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](N)Cc8c

InChI

1S/C157H232N40O47S2/c1-79(2)57-104(181-130(211)86(15)172-136(217)103(50-53-125(209)210)180-152(233)127(84(11)12)194-148(229)107(60-82(7)8)184-145(226)113(67-95-70-165-78-171-95)189-150(231)115(74-198)176-123(206)73-169-133(214)116(75-245(239,240)241)191-140(221)105(58-80(3)4)182-144(225)112(66-94-69-164-77-170-94)188-138(219)102(48-51-119(160)202)178-146(227)114(68-120(161)203)190-153(234)126(83(9)10)193-131(212)98(159)61-89-31-21-18-22-32-89)139(220)185-110(64-92-40-44-96(200)45-41-92)142(223)183-106(59-81(5)6)147(228)195-128(85(13)14)154(235)192-117(76-246(242,243)244)134(215)168-71-121(204)174-101(49-52-124(207)208)137(218)177-99(38-29-55-166-157(162)163)132(213)167-72-122(205)175-108(62-90-33-23-19-24-34-90)141(222)186-109(63-91-35-25-20-26-36-91)143(224)187-111(65-93-42-46-97(201)47-43-93)149(230)196-129(88(17)199)155(236)197-56-30-39-118(197)151(232)179-100(37-27-28-54-158)135(216)173-87(16)156(237)238/h18-26,31-36,40-47,69-70,77-88,98-118,126-129,198-201H,27-30,37-39,48-68,71-76,158-159H2,1-17H3,(H2,160,202)(H2,161,203)(H,164,170)(H,165,171)(H,167,213)(H,168,215)(H,169,214)(H,172,217)(H,173,216)(H,174,204)(H,175,205)(H,176,206)(H,177,218)(H,178,227)(H,179,232)(H,180,233)(H,181,211)(H,182,225)(H,183,223)(H,184,226)(H,185,220)(H,186,222)(H,187,224)(H,188,219)(H,189,231)(H,190,234)(H,191,221)(H,192,235)(H,193,212)(H,194,229)(H,195,228)(H,196,230)(H,207,208)(H,209,210)(H,237,238)(H4,162,163,166)(H,239,240,241)(H,242,243,244)/t86-,87-,88+,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,108-,109-,110-,111-,112-,113-,114-,115-,116-,117-,118-,126-,127-,128-,129-/m0/s1

InChI key

CZJQESISDHGFTC-DQCCOYHDSA-N

Gene Information

cow ... INS(280829)

Biochem/physiol Actions

Insulin regulates the cellular uptake, utilization, and storage of glucose, amino acids, and fatty acids. It inhibits the breakdown of glycogen, protein, and fat. The β chain is a substrate for carboxypeptidase Y.

Preparation Note

HPLC-purified and use-tested as a protease substrate.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
I6383I1633I6634
product line

BioReagent

product line

-

product line

-

product line

BioReagent

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

technique(s)

protein sequencing: suitable

technique(s)

-

technique(s)

-

technique(s)

cell culture | mammalian: suitable, single cell analysis: suitable

Gene Information

cow ... INS(280829)

Gene Information

cow ... INS(280829)

Gene Information

cow ... INS(280829)

Gene Information

cow ... INS(280829)

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

UniProt accession no.

P01317

UniProt accession no.

P01317

UniProt accession no.

P01317

UniProt accession no.

P01317


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters