Skip to Content
MilliporeSigma
All Photos(1)

Documents

HT100A

Sigma-Aldrich

Silver Stain (Modified GMS) Kit

Synonym(s):

Modified Grocott Methanamine Silver (GMS) Fungus Stain, Periodic Acid Methanamine Silver Technique

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116124
NACRES:
NA.47

form

solution

shelf life

Expiry date on the label

IVD

for in vitro diagnostic use

application(s)

hematology
histology

storage temp.

2-8°C

Diagnostic Implications

The Silver Stain modified GMS kit is intended for use in the histologic demonstration of fungi,basement membrane and some opportunistic organisms. The Silver Stain kit is for “In VitroDiagnostic Use.” For professional use only. The data obtained from this manual, qualitativeprocedure identifies fungi, basement membrane and some opportunistic organisms in tissuesamples of human specimens. This data when reviewed in conjunction with other diagnostic testsand information may be used as an aid to diagnosis of fungal infections.

Kit Components Only

Product No.
Description

  • Borax Solution (HT100-2) 100 mL

  • Gold Chloride Solution (HT100-4) 100 mL

  • Periodic Acid Solution (kit only) 100 mL

  • Silver Methenamine Reagent (HT100-3) 12 vial(s)

  • Sodium Thiosulfate Solution (HT100-5) 100 mL

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Ox. Liq. 1 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Thyroid

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tereza Juříková et al.
Journal of fungi (Basel, Switzerland), 6(4) (2020-11-05)
A procedure for processing frozen rat lung tissue sections for scanning electron microscopy (SEM) from deeply frozen samples initially collected and stored for matrix-assisted laser desorption/ionization mass spectrometry imaging (MALDI-MSI) was developed. The procedure employed slow thawing of the frozen
Chung-Ping Liao et al.
Antimicrobial agents and chemotherapy, 53(12), 5259-5264 (2009-10-07)
Polyamine levels are greatly increased in alveolar macrophages (AMs) during Pneumocystis pneumonia (PCP), leading to increased production of H(2)O(2), which causes AMs to undergo apoptosis. One of the mechanisms by which polyamine levels in AMs are elevated is enhanced uptake
Genki Kimura et al.
Antimicrobial agents and chemotherapy, 61(9) (2017-06-21)
PC945 is a novel triazole optimized for lung delivery, and the objective of this study is to determine the effects of intranasally dosed PC945 on Aspergillus fumigatus infection and associated biomarkers in immunocompromised mice. PC945, posaconazole, or voriconazole was administered
Genki Kimura et al.
Mycoses, 60(11), 728-735 (2017-07-13)
Although anti-fungal triazoles are dosed orally or systemically for Aspergillus fumigatus infection, systemic adverse events and limited exposure of the lung cavity would make a topical treatment for the lung an attractive option. In this study, we examined the effects
Yinqiu Wang et al.
Journal of the American Society of Nephrology : JASN, 21(10), 1657-1666 (2010-07-24)
Mutant forms of TRPC6 can activate NFAT-dependent transcription in vitro via calcium influx and activation of calcineurin. The same TRPC6 mutants can cause FSGS, but whether this involves an NFAT-dependent mechanism is unknown. Here, we generated mice that allow conditional

Articles

Fungal stains remain an important tool in the histology laboratory's diagnostic arsenal for identifying infectious microorganisms.

Fungal stains remain an important tool in the histology laboratory's diagnostic arsenal for identifying infectious microorganisms.

Fungal stains remain an important tool in the histology laboratory's diagnostic arsenal for identifying infectious microorganisms.

Fungal stains remain an important tool in the histology laboratory's diagnostic arsenal for identifying infectious microorganisms.

Related Content

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service