Documents
HS23381B
Arctic Squares® Cryostorage Box
Holds 81 x 2 mL tubes, assorted colors polycarbonate, pk of 4
About This Item
UNSPSC Code:
41123304
NACRES:
NB.22
material
assorted colors polycarbonate
feature
Holds 81 x 2 mL tubes
packaging
pk of 4
manufacturer/tradename
Heathrow Scientific HS23381B
technique(s)
cryopreservation: suitable
size
133x133x53 mm
General description
Safely stores vials from -196 to 121C in mechanical freezers as well as liquid nitorgen. Boxes are designed with unique features to help orient, identify and access vials with ease.
  • forward sloped base and high contrast indexing on the transparent lid ensure quick visual orientation
  • vend and drainage holes
  • stackable
  • autoclavable
Features and Benefits
  • hinged lid with built-in stop, allows easy one-handed access to samples
  • wells arranged in ans easy-to-pick formation
  • magnetic tipped forceps supplied to help pick vials and remove boxes from freezer
  • ideal for small sampling
  • friction fit, slide on lid
Legal Information
Arctic Squares is a registered trademark of Heathrow Scientific LLC