Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA031469

Sigma-Aldrich

Anti-HRNR antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-S100A16, Anti-S100a18, Anti-hornerin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

WSAGENDSYSRNVRGSLKPGTESISRRLSFQRDFSGQHNSYSGQSSSYGEQNSDSHQSSGRGQCGSGS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HRNR(388697)

General description

The gene HRNR (hornerin) is mapped to human chromosome 1q21. It belongs to the S100 protein family and is a calcium binding protein. The protein has a Ca2+ binding EF-hand domain and a repetitive domain rich in glycine and serine.

Immunogen

hornerin recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-HRNR antibody produced in rabbit has been used in immunofluorescence and western blotting.

Biochem/physiol Actions

HRNR (hornerin) might be involved in keratinocyte cornification. It is also expressed in breast tissue and its expression changes according to breast cancer subtype. Induction of apoptosis or necrosis upregulates the expression of HRNR in breast cells. Mutation in this gene is associated with risk of atopic dermatitis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77200

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chromosome 11q13.5 variant associated with childhood eczema: an effect supplementary to filaggrin mutations.
O'Regan GM, et al.
The Journal of Allergy and Clinical Immunology, 125, 170-174 (2010)
Laurence Pellerin et al.
The Journal of allergy and clinical immunology, 131(4), 1094-1102 (2013-02-14)
Atopic dermatitis (AD) is a chronic inflammatory skin disease characterized by a disturbed epidermal barrier. In a subset of patients, this is explained by nonsense mutations in the gene encoding filaggrin (FLG). We sought to evaluate the respective role of
Julie Henry et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 25(5), 1567-1576 (2011-02-02)
A single-nucleotide polymorphism within the gene encoding hornerin (HRNR) has recently been linked with atopic dermatitis (AD) susceptibility. HRNR shares features with filaggrin, a key protein for keratinocyte differentiation, but conflicting reports have been published concerning its expression in the
Jodie M Fleming et al.
BMC cancer, 12, 266-266 (2012-06-26)
Recent evidence suggests an emerging role for S100 protein in breast cancer and tumor progression. These ubiquitous proteins are involved in numerous normal and pathological cell functions including inflammatory and immune responses, Ca(2+) homeostasis, the dynamics of cytoskeleton constituents, as
Michael F Gutknecht et al.
Nature communications, 8(1), 552-552 (2017-09-17)
Sustained angiogenesis is essential for the development of solid tumors and metastatic disease. Disruption of signaling pathways that govern tumor vascularity provide a potential avenue to thwart cancer progression. Through phage display-based functional proteomics, immunohistochemical analysis of human pancreatic ductal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service