Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA020138

Sigma-Aldrich

Anti-SVIL antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-Archvillin, Anti-Supervillin, Anti-p205/p250

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

NARYQTQPVTLGEVEQVQSGKLIPFSPAVNTSVSTVASTVAPMYAGDLRTKPPLDHNASATDYKFSSSIENSDSPVRSILKSQAWQPLVEGSENKGMLREYGETESKRALTGRDSGMEKYGSFEEAEASYPILNRARE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SVIL(6840)

General description

Supervillin (SVIL) is a 205kDa protein expressed in the peripheral membrane. It possesses an amino-terminal domain and a carboxyl-terminal domain containing a villin-type domain. The gene encoding SVIL is localized on human chromosome 10p11.2.

Immunogen

Supervillin recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Supervillin (SVIL) binds to F-actin, myosin II and other signaling proteins in lipid rafts. It accelerates the signaling pathways from the epidermal growth factor receptor (EGFR) to extracellular signal-regulated kinases (ERKs). Thus, it coordinates with many proteins to help in cell motility. SVIL increases the contractile nature of myosin II by scaffolding it to long-myosin light chain kinase (L-MLCK). It also contributes to enhance extracellular matrix degradation and reducing the extent of cell-substrate adhesion. SVIL expression in cells have negative effects on thrombus development.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74722

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R K Pope et al.
Genomics, 52(3), 342-351 (1998-12-29)
Supervillin is a 205-kDa F-actin binding protein originally isolated from bovine neutrophils. This protein is tightly associated with both actin filaments and plasma membranes, suggesting that it forms a high-affinity link between the actin cytoskeleton and the membrane. Human supervillin
Zhiyou Fang et al.
Traffic (Copenhagen, Denmark), 11(6), 782-799 (2010-03-25)
In migrating cells, the cytoskeleton coordinates signal transduction and redistribution of transmembrane proteins, including integrins and growth factor receptors. Supervillin is an F-actin- and myosin II-binding protein that tightly associates with signaling proteins in cholesterol-rich, 'lipid raft' membrane microdomains. We
Jeffrey W Brown et al.
Journal of molecular biology, 393(3), 608-618 (2009-08-18)
Villin-type headpiece domains are compact motifs that have been used extensively as model systems for protein folding. Although the majority of headpiece domains bind actin, there are some that lack this activity. Here, we present the first NMR solution structure
Leonard C Edelstein et al.
Circulation, 125(22), 2762-2771 (2012-05-03)
High shear force critically regulates platelet adhesion and thrombus formation during ischemic vascular events. To identify genetic factors that influence platelet thrombus formation under high shear stress, we performed a genome-wide association study and confirmatory experiments in human and animal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service