Skip to Content
MilliporeSigma
All Photos(8)

Key Documents

HPA019039

Sigma-Aldrich

Anti-TOP1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DNA topoisomerase 1, Anti-DNA topoisomerase I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, rat, mouse

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

KVRASGDAKIKKEKENGFSSPPQIKDEPEDDGYFVPPKEDIKPLKRPRDEDDADYKPKK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TOP1(7150)

General description

The gene TOP1 (DNA topoisomerase 1) is mapped to human chromosome 20q12-q13.1. The protein localizes in the nucleus.

Immunogen

DNA topoisomerase 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

DNA topoisomerases cause single (type-1) or double (type-2) strand DNA breaks, leading to DNA relaxation. They control DNA replication, transcription, recombination and chromatin remodeling. TOP1 (DNA topoisomerase 1) binding to the promoter areas induces nucleosome disassembly, histone acetylation and gene expression. Camptothecin is a non-competitive TOP1 inhibitor. TOP1 also controls alternative splicing through phosphorylation of serine/arginine-rich proteins. It is up-regulated in non-small cell lung cancer (NSCLC).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74819

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshiaki Onishi et al.
Nucleic acids research, 40(19), 9482-9492 (2012-08-21)
The Bmal1 gene is essential for the circadian system, and its promoter has a unique open chromatin structure. We examined the mechanism of topoisomerase I (Top1) to understand the role of the unique chromatin structure in Bmal1 gene regulation. Camptothecin
Ann-Katrine Jakobsen et al.
Experimental and molecular pathology, 99(1), 56-64 (2015-05-20)
Topoisomerase I (TOP1) regulates DNA topology during replication and transcription whereas tyrosyl-DNA phosphodiesterase 1 (TDP1) is involved in the repair of several types of DNA damages, including damages from defective TOP1 catalysis. TOP1 is the target of chemotherapeutic drugs of
Steffen Israel et al.
Scientific reports, 9(1), 13356-13356 (2019-09-19)
Early mouse embryos have an atypical translational machinery that consists of cytoplasmic lattices and is poorly competent for translation. Hence, the impact of transcriptomic changes on the operational level of proteins is predicted to be relatively modest. To investigate this
Chen Zhao et al.
Cancer, 98(1), 18-23 (2003-07-02)
Amplification of DNA in certain chromosomal regions plays a crucial role in the development and progression of human malignancies, specifically when protooncogenic target genes within those amplicons are overexpressed. Comparative genomic hybridization studies have revealed frequent amplification at 20q in
Andreas Eisenreich et al.
Circulation research, 104(5), 589-599 (2009-01-27)
Tumor necrosis factor (TNF)-alpha-stimulated human umbilical vein endothelial cells express 2 naturally occurring forms of tissue factor (TF), the primary initiator of blood coagulation: the soluble alternatively spliced isoform and the full-length TF isoform. The regulatory pathways enabling this phenomenon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service