Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

HPA010088

Sigma-Aldrich

Anti-CD63 antibody produced in rabbit

affinity isolated antibody, Prestige Antibodies® Powered by Atlas Antibodies, buffered aqueous glycerol solution

Synonym(s):

Anti-CD63 antigen, Anti-Granulophysin, Anti-Lamp-3, Anti-Lysosomal-associated membrane protein 3, Anti-Melanoma-associated antigen ME491, Anti-OMA81H, Anti-Ocular melanoma- associated antigen, Anti-Tetraspanin-30, Anti-Tspan-30

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:500-1:1000

immunogen sequence

RDKVMSEFNNNFRQQMENYPKNNHTASILDRMQADFKCCGAANYTDWEKIPSMSKNRVPDSCCINVTVGCGINFNEKAIHKEGCVEKIG

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD63(967)

General description

CD63 (cluster of differentiation) is a membrane protein belonging to the tetraspanin family of proteins called LAMP (lysosome-associated membrane glycoprotein) proteins. It is localized to histamine-containing granules.
It also resides in MHC (major histocompatibility compartment) II endocytic compartment.

Immunogen

CD63 antigen recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CD63 (cluster of differentiation) is associated with neutrophil homeostasis in a TIMP-1 (tissue inhibitor of metalloproteinases-1)-mediated manner. This protein is also involved in promoting metastasis, through the stabilization of β-catenin. It also affects the plasticity of cell. Its expression is high during early stages of melanoma, which decreases with the progression of melanoma. Thus, it functions as a tumor suppressor by inhibiting EMT (epithelial-mesenchymal transition) in melanoma. CD63, along with CD9, prevents cell-cell fusion in HIV (human immunodeficiency virus) infection, during transition from hemifusion to pore opening.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72477

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra Pinet et al.
Oncotarget, 7(31), 50349-50364 (2016-07-08)
The neurotrophin receptors are known to promote growth and proliferation of glioblastoma cells. Their functions in spreading glioblastoma cell aggressiveness to the microenvironment through exosome release from glioblastoma cells are unknown. Considering previous reports demonstrating that YKL-40 expression is associated
Takahiro Kishikawa et al.
JCI insight, 1(8), e86646-e86646 (2016-10-05)
Pancreatic ductal adenocarcinoma (Pdac) is a malignancy with a poor prognosis due to difficulties in early detection. Although promising biomarkers are increasingly reported, such methods are not yet easy to apply clinically, mainly due to their low reproducibility or technical
Julia Kobuch et al.
Haematologica, 100(8), 1005-1013 (2015-05-24)
The homeostasis of neutrophil granulocytes can affect the outcome of several inflammation-associated diseases including cancer. The regulation of this homeostasis is still not completely understood. We previously found that elevated systemic levels of tissue inhibitor of metalloproteinases-1 (TIMP-1) induce an
Bastian Seubert et al.
International journal of cancer, 136(10), 2304-2315 (2014-10-30)
The tetraspanin CD63 is implicated in pro-metastatic signaling pathways but, so far, it is unclear, how CD63 levels affect the tumor cell phenotype. Here, we investigated the effect of CD63 modulation in different metastatic tumor cell lines. In vitro, knock
Shin La Shu et al.
Methods in molecular biology (Clifton, N.J.), 2265, 289-304 (2021-03-12)
Cells release extracellular vesicles (EVs) that can be detected both in vivo and in cell culture medium. Among EVs, exosomes are 50-150 nm vesicles that are systematically packaged into multivesicular bodies for release into the external environment. In cancer, these intentionally

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service