Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

HPA002642

Sigma-Aldrich

Anti-ITGA5 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

ITGA5 Antibody - Anti-ITGA5 antibody produced in rabbit, Itga5 Antibody, Anti-CD49e antigen) [Contains: Integrin α-5 heavy chain; Integrin α-5 light chain] antibody produced in rabbit, Anti-Fibronectin receptor subunit α antibody produced in rabbit, Anti-Integrin α-5 precursor antibody produced in rabbit, Anti-Integrin α-F antibody produced in rabbit, Anti-VLA-5 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, mouse

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

DQPQKEEDLGPAVHHVYELINQGPSSISQGVLELSCPQALEGQQLLYVTRVTGLNCTTNHPINPKGLELDPEGSLHHQQKREAPSRSSASSGPQILKCPEAECFRLRCELGPLHQQESQSLQLHFRVWA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ITGA5(3678)

General description

ITGA5 (integrin subunit α 5) is a transmembrane glycoprotein. The protein has a large extracellular domain, a single spanning transmembrane domain and a short cytoplasmic domain.

Immunogen

Integrin α-5 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-ITGA5 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Broadly, integrins are associated with adhesive interactions between the trophoblast (blastocyst) and uterine epithelium at implantation. The ITGA5 (integrin subunit α 5) associates during dendritic cell (DC) trafficking with vascular-cell adhesion molecule-1 to mediate pre DC2 (Dendritic-cell) adhesion/transmigration. During normal hemopoiesis, cytokine-dependent activation of ITGA5 acceleartes the human hemopoietic cell adhesiveness.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86565

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Zou et al.
Nature medicine, 7(12), 1339-1346 (2001-12-01)
Dendritic-cell (DC) trafficking and function in tumors is poorly characterized, with studies confined to myeloid DCs (DC1s). Tumors inhibit DC1 migration and function, likely hindering specific immunity. The role of plasmacytoid DCs (DC2s) in tumor immunity is unknown. We show
J P Lévesque et al.
The Journal of experimental medicine, 181(5), 1805-1815 (1995-05-01)
Cytokines are known to be important regulators of normal hemopoiesis, acting in concert with components of the bone marrow microenvironment. Interactions with this microenvironment are known to regulate the proliferation, differentiation, and homing of hemopoietic progenitor (CD34+) cells. Adhesive interactions
Sarah Paule et al.
Human reproduction (Oxford, England), 27(9), 2766-2774 (2012-06-29)
Proprotein convertases (PCs) post-translationally activate a large number of protein precursors through limited cleavage. PC5/6 (PC6) in the human endometrium is tightly regulated during receptivity for embryo implantation. Integrins are transmembrane glycoproteins, some of which play an important role in
Jodi A McKenzie et al.
Carcinogenesis, 34(9), 2137-2144 (2013-05-04)
Survivin is an apoptotic and mitotic regulator that is overexpressed in melanoma and a poor prognostic marker in patients with metastatic disease. We recently showed that Survivin enhances melanoma cell motility through Akt-dependent upregulation of α5 integrin. However, the functional
Ling Lu et al.
Molecular oncology, 13(12), 2697-2714 (2019-10-11)
The tumorigenesis of colorectal cancer (CRC) is a complicated process, involving interactions between cancer cells and the microenvironment. The role of α5 integrin subunit in CRC remains controversial, and previous studies mainly focused on cancer cells. Herein, we report an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service