Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GS85

Sigma-Aldrich

GST A1-1, Recombinant Bovine

Synonym(s):

glutathione S-transferase alpha 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

biological source

bovine

Quality Level

recombinant

expressed in E. coli

Assay

>95% (SDS-PAGE)

form

frozen liquid

specific activity

4.35 units/mg protein

mol wt

23 kDa

concentration

1.42 mg/mL

storage temp.

−70°C

General description

using spectrophotometric determination of 1-chloro-2,4-dinitrobenzene (CDNB) conjugation with reduced glutathione (1 mM) in 100 mM NaPO4 (pH 6.5) at room temperature.

Storage and Stability

The enzyme should be used by the end-user customer within 1 year of receipt.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Rabahi et al.
Endocrinology, 140(8), 3507-3517 (1999-08-05)
We have previously shown that a major group of 28-30 kDa proteins decreases after the LH surge in bovine granulosa cells (GC). In the present study, we have characterized two proteins in this group in search of factors that may

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service