Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE80-1129-53

EPH electrode wick for MultiPhor II electrophoresis unit

W × L 82 mm × 130 mm, pack of 500 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105300
NACRES:
NB.22

grade

electrophoresis grade

packaging

pack of 500 ea

manufacturer/tradename

Cytiva 80112953

W × L

82 mm × 130 mm

Looking for similar products? Visit Product Comparison Guide

General description

Used for connecting the buffer solution with the gel.

Legal Information

MultiPhor is a trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service