Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE27-0361-01

Ready-to-go T4 DNA Ligase

Cytiva 27-0361-01, sufficient for 50 reactions

Synonym(s):

T4 DNA ligase enzyme

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.53

usage

sufficient for 50 reactions

manufacturer/tradename

Cytiva 27-0361-01

General description

Ready-To-Go T4 DNA Ligase. Catalyzes the formation of a phosphodiester bond between the 5′-phosphoryl group and the 3′-hydroxyl group of two double-stranded DNA fragments. ATP is required for this reaction. Ligation of blunt-ended DNA is greatly stimulated by polyethylene glycol.

Application

Applications include self-circularization of linear DNA prior to transformation, ligation of insert to vector, and ligation of synthetic linkers to blunt-ended DNA. Each predispensed, single-reaction, ambient-temperature-stable tube contains a glassified reaction mix which, when reconstituted to a final reaction volume of 20 μL, yields a solution of reaction buffer, T4 DNA ligase, and ATP.

Features and Benefits

  • For covalently-linking DNA fragments in as little as 30 min.
  • Single-dose, ambient-temperature-stable reaction mixes.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Ready-To-Go is a trademark of Cytiva

related product

Storage Class Code

13 - Non Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service