Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17371501

Capto Adhere ImpRes, 25 ML

Cytiva 17371501, 40 μm avg. part. size (median particle size of the cumulative volume distribution)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47101511
NACRES:
NA.56

ligand

(Multimodal strong anion exchanger)

manufacturer/tradename

Cytiva 17371501

matrix

high-flow agarose

avg. part. size

40 μm (median particle size of the cumulative volume distribution)

cleaning in place

2-14

working range

3-12

capacity

45-85 mg Mab/ml binding capacity (Chromatography Medium)

General description

The multimodal anion exchanger ligand of Capto adhere ImpRes is immobilized to the base matrix and interacts with the target molecule through multiple types of interactions.

The high selectivity of the anion multimodal ligand allows operation in a variety of process conditions to solve challenging purification problems. Contaminants such as DNA, host cell proteins, leached protein A, aggregates, and viruses are efficiently removed.

Capto adhere ImpRes is available in a range of different bulk pack sizes for easy scale-up and pre-packed formats for process development.

As member of the BioProcess media range, Capto adhere ImpRes meets industrial demands with security of supply and comprehensive technical and regulatory support.

Features and Benefits

Capto adhere ImpRes is a strong anion exchange multimodal BioProcess chromatography medium (resin), designed to meet the demands of modern large-scale manufacturers, for high-resolution polishing of monoclonal antibodies (MAbs) and other biomolecules.

  • High yields achieved through the high-resolution beads and selectivity of the ligand.
  • Efficient removal of aggregates, viruses, and main contaminants in MAb processes.
  • Enables use of a platform approach to MAb process development.
  • Allows separation of MAb charged variants.

Storage and Stability

Store at 4 to 30 °C (20% ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

In these studies, the binding capacity for MAbs and the efficiency in the clearance of impurities using Capto adhere ImpRes in bind/elute mode was evaluated.

This page describes the maintenance of media and storage conditions for multimodal chromatography using Cytiva products.

Protocols

Capto adhere ImpRes is a cost-effective and flexible chromatography medium from Cytiva, designed for high-resolution polishing of MAbs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service