Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-0569-01

EAH-Sepharose 4B

Cytiva 17-0569-01, pack of 50 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

packaging

pack of 50 mL

manufacturer/tradename

Cytiva 17-0569-01

storage condition

(20% Ethanol containing 0.9% NaCl)

matrix

4% agarose

particle size

45-165 μm

average diameter

90 μm

ligand dependent

3-14

storage temp.

2-8°C

General description

EAH Sepharose pre-activated media is used for coupling compounds containing carboxyl groups to Sepharose 4B through carbodiimide- based coupling via an 11-atom spacer arm.

Application

Convenient immobilization of ligands using pre-activated medium.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows how to convert between flow velocity and volumetric flow rate in affinity chromatography of antibodies.

Protocols

Ligands are coupled in a simple one-step procedure in the presence of a coupling reagent, carbodiimide. The carbodiimides may be regarded as anhydrides of urea.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service