Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-0440-01

Con-A Sepharose 4B

Cytiva 17-0440-01, pack of 100 mL

Synonym(s):

Concanavalin A Sepharose

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

ligand

concanavalin A (con A)

packaging

pack of 100 mL

manufacturer/tradename

Cytiva 17-0440-01

storage condition

(0.1 M acetate buffer pH 6, 1 M NaCl, 1 mM CaCl2, 1 mM MnCl2 and 1 mM MgCl, 20% Ethanol)

matrix

4% agarose

particle size

45-165 μm

average diameter

90 μm

cleaning in place

4-9

working range

4-6

storage temp.

2-8°C

General description

Con A Sepharose 4B with immobilized Concanavalin A is an affinity chromatography media for capture of glycosylated biomolecules including glycoproteins and glycolipids. Stronger affinity for α-D-glucose than α-D-mannose.

Con A Sepharose is Concanavalin A coupled to Sepharose 4B by the cyanogen bromide method. Concanavalin A (Con A) is a tetrameric metalloprotein isolated from Canavalia ensiformis (jack bean). Con A binds molcules containing α-D-mannopyranosyl, α-D-glucopyranosyl and sterically related residues. The binding sugar requires the presence of C-3, C-4 and C-5 hydroxyl groups for reaction with Con A. Con A coupled to Sepharose is routinely used for separation and purification of glycoproteins, polysaccharides and glycolipids.

Features and Benefits

  • Concanavalin A (Con A) binds molecules that contain α-D-mannose, α-D-glucose, and sterically related residues with available C-3, C-4, or C-5 hydroxyl groups
  • Group-specific adsorbent for molecules containing sugars.
  • Highly suitable for isolation of cell surface glycoproteins from detergent-solubilized membranes
  • Concanavalin A (Con A) binds molecules that contain α-D-mannose, α-D-glucose and sterically related residues with available C-3, C-4, or C-5 hydroxyl groups
  • Highly suitable for isolation of cell surface glycoproteins from detergent-solubilized membranes
  • Con A coupled to Sepharose 4B via cyanogen bromide activation

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows how to convert between flow velocity and volumetric flow rate in affinity chromatography of antibodies.

This page shows how to solve practical problems that may occur when running an affinity chromatography column.

Protocols

This page shows how to purify or remove glycoproteins and polysaccharides with Con A Sepharose 4B, Lentil Lectin Sepharose 4B, Capto Lentil Lectin from Cytiva.

This page shows how to perform column packing and preparation for affinity chromatography when using Tricorn™ or XK columns available from Cytiva.

This page shows how to prepare samples for purification with affinity chromatography.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service