Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE10600116

Amersham Protran® Western blotting membrane sandwich, nitrocellulose

pore size 0.1 μm, sheet W × L 80 mm × 90 mm , preassembled with 2 x 3MM chr filter papers, pkg of 10 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
NACRES:
NB.22

packaging

pkg of 10 ea

manufacturer/tradename

Cytiva 10600116

sheet W × L

80 mm × 90 mm , preassembled with 2 x 3MM chr filter papers

pore size

0.1 μm pore size

Looking for similar products? Visit Product Comparison Guide

General description

Amersham Protran® NC Sandwich contains 10 Western blotting sandwiches, consisting of precut Amersham Protran® nitrocellulose membranes preassembled with 2 x 3MM Chr filter papers. Designed for convenience and ease-of-use.

Features and Benefits

  • Precut Amersham Protran 0.1 µm nitrocellulose membranes.
  • Sandwich packs are designed for convenience and ease of use.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

Amersham is a trademark of Cytiva
Protran is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service