Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE10600046

Amersham Protran® Western blotting membranes, nitrocellulose

pore size 0.1 μm, sheet W × L 200 mm × 200 mm, pkg of 10 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
NACRES:
NB.22

packaging

pkg of 10 ea

manufacturer/tradename

Cytiva 10600046

sheet W × L

200 mm × 200 mm

pore size

0.1 μm pore size

Looking for similar products? Visit Product Comparison Guide

General description

A versatile nitrocellulose blotting membrane with 0.1 µm pore size. Amersham Protran offers excellent sensitivity, resolution and low background, especially when used with Amersham ECL, Amersham ECL Prime, or Amersham ECL Select Western blotting systems.

Application

Amersham Protran® Western blotting membranes, nitrocellulose is suitable for use in:
  • Western blotting
  • Southern blotting
  • Northern blotting

Features and Benefits

  • Low background and excellent signal-to-noise ratios.
  • Compatible with a variety of labeling and detection systems.
  • High binding capacity of small proteins (< Mr 10 000) reduces risk of "blow-through".
  • Proprietary nitrocellulose formula gives a long shelf life of bound proteins.
  • Available in popular precut sizes for laboratory application.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

Amersham is a trademark of Cytiva
Protran is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service