Key Documents
GBL665301
Grace Bio-Labs Press-To-Seal silicone isolator, PSA one side
(Adhesive on one side)
Synonym(s):
Nonadhesive reagent isolators, autoclavable slide chambers, removable hydrophobic barrier
About This Item
UNSPSC Code:
41161502
NACRES:
NB.13
material
red silicon
feature
(Adhesive on one side)
packaging
pack of 25 ea
manufacturer/tradename
Grace Bio-Labs 665301
external L × W
51 mm × 25 mm
well area
0.66 cm2
wells
8
General description
Silicone Isolators allow researchers to isolate specimens using removable hydrophobic barriers. They may be used to isolate cells grown in culture dishes or separate multiple specimens affixed to microscope slides. Isolators are available in red silicone thickness 0.5 to 2.5mm pre-cut or as sheet material with or without secure seal clean adhesive on one or both surfaces. Nonadhesive isolators are autoclavable and reusable.
Silicone Isolators remain sealed to smooth surfaces during washing steps. Where additional sealing is required, SecureSeal adhesive on one or both surfaces is recommended. Each seal design is also available in CultureWell "water-clear" silicone, which is highly recommended for sensitive cell culture applications. Closed chambers may be formed using flexible RNase free, HybriSlip covers. Recommended use temperature is -62°C to 218°C.