Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EPI010

Sigma-Aldrich

Sirtuin 2 (SIRT2) Inhibitor Screening Assay Kit

100 assays in 96 well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.41

usage

100 assays in 96 well plates

storage condition

protect from light

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SIRT2(22933)
mouse ... SIRT2(64383)

General description

Sirtuin 2 (SIRT2) is a cytoplasmic protein and is part of the sirtuin family of proteins. Sirtuin family contains a sirtuin core domain and isgrouped into four classes with SIRT2 being a member of class I. SIRT2 colocalizes with microtubules. The gene encoding this protein is localized on human chromosome 19q13.2.

With Sigma′s Sirtuin Inhibitor Screening Kit, Sirtuin deacetylates the Substrate, and then a Developer cleaves the deacetylated substrate to release a fluorescent group, the latter of which can be detected at Ex/Em = 395/541 nm. In the presence of a SIRT inhibitor, the deacetylation will be impeded, preventing substrate cleavage and release of the fluorescent group. The kit provides a rapid, simple, sensitive, and reliable test, which is suitable for both low and high-throughput screening of SIRT2 inhibitors. A positive control inhibitor, Nicotinamide, is included to compare with the efficacy of the test inhibitors.

Application

Sirtuin 2 (SIRT2) inhibitor screening assay kit has been used for acetyl peptide deacetylase assay.

Biochem/physiol Actions

Sirtuin 2 (SIRT2) maintains the integrity of the genome. Inhibition of SIRT2 can lead to neuroprotection in cellular and invertebrate models of Huntington′s disease. Huntington′s disease is characterized by increased sterol synthesis in neuronal cells and this process is reversed by SIRT2 inhibition. SIRT2 can deacetylate lys40 of α-tubulin both in vitro and in vivo. Knockdown of SIRT2 by small interfering RNA (siRNA) leads to tubulin hyperacetylation.

Features and Benefits

  • Simple, sensitive, and reliable assay
  • Species reactivity: mammalian
  • Utilizes fluorometric methods
  • Suitable for individual tests or high throughput assays
  • Convenient 96-well microplate format
  • Screening and characterization of Sirtuin inhibitors

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The human Sir2 ortholog, SIRT2, is an NAD+-dependent tubulin deacetylase.
North BJ
Molecular and Cellular Biochemistry, 11(2) (2003)
Damir Janigro
The Cell Cycle in the Central Nervous System (2008)
Brian P Weiser et al.
The Journal of biological chemistry, 290(13), 8559-8568 (2015-02-11)
meta-Azi-propofol (AziPm) is a photoactive analog of the general anesthetic propofol. We photolabeled a myelin-enriched fraction from rat brain with [(3)H]AziPm and identified the sirtuin deacetylase SIRT2 as a target of the anesthetic. AziPm photolabeled three SIRT2 residues (Tyr(139), Phe(190)
Propofol Inhibits SIRT2 Deacetylase through a Conformation-specific, Allosteric Site*
Brian P
The Journal of Biological Chemistry (2015)
SIRT2 inhibition achieves neuroprotection by decreasing sterol biosynthesis
Ruth Luthi-Carter
Proceedings of the National Academy of Sciences of the USA (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service