Key Documents
EP3125000125
Eppendorf Research® Plus Move It pipette
no. of channels, 12, volume 0.1-20 μL, 4.5-9 mm cone distance, light pink
Synonym(s):
Eppendorf® Research® Plus Move It pipette, Pipettor, multichannel pipette, multichannel pipettor
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
material
light pink
feature
adjustable
manual
no. of channels 12
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 3125000125
volume
0.1-20 μL
General description
Eppendorf® Research plus Move It, 12-channel, 4.5-9 mm cone distance, variable, incl. epT.I.P.S.® Box 2.0, 1 – 20 µL, light pin
Features and Benefits
  • 4-, 6-, 8- and 12-channel air-cushion pipettes with variable volume and adjustable tip spacing option
  • Available as mechanical (Research plus) and electronic (Xplorer plus) adjustable tip spacing multi-channel pipette
  • For plates (384-, 96-, 48-, 24- and 12-well), tubes, agarose gels and more
  • Tip spacing freely selectable from 4.5 to 33 mm, depending on pipette variant
  • Format limiter for quick switches backwards and forwards between source and destination formats - Manual adjustment knob for smooth vibration-free format switches and liquid transfers without dripping
  • Tubeless system for enhanced reliability and safety - Autoclavable for user and sample safety (Research plus fully, and Xplorer plus lower part only)
  • 360° rotatable pipette head for comfortable readability of display and fast identification of parameters
  • Optimally balanced pipette - reduces hand fatigue for stress-free operation.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Research is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG