Skip to Content
MilliporeSigma
All Photos(1)

Documents

EHU070231

Sigma-Aldrich

MISSION® esiRNA

targeting human SLC7A11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ATCTTTGTTGCCCTCTCCTGCTTTGGCTCCATGAACGGTGGTGTGTTTGCTGTCTCCAGGTTATTCTATGTTGCGTCTCGAGAGGGTCACCTTCCAGAAATCCTCTCCATGATTCATGTCCGCAAGCACACTCCTCTACCAGCTGTTATTGTTTTGCACCCTTTGACAATGATAATGCTCTTCTCTGGAGACCTCGACAGTCTTTTGAATTTCCTCAGTTTTGCCAGGTGGCTTTTTATTGGGCTGGCAGTTGCTGGGCTGATTTATCTTCGATACAAATGCCCAGATATGCATCGTCCTTTCAAGGTGCCACTGTTCATCCCAGCTTTGTTTTCCTTCACATGCCTCTTCATGGTTGCCCTTTCCCTCTATTCGGACCCATTTAGTACAGGGATTGGCTTCGTCATCACT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naisu Yang et al.
Journal of genetics, 97(2), 463-468 (2018-06-23)
Solute carrier family 7 member 11 (SLC7A11) is a cystine/glutamate exchanger, also known as xCT, has been found to play an important role in pheomelanin synthesis. Adjusting the cystine content of cells to influence pheomelanin synthesis affects the proportion of
Masaki Nagane et al.
PloS one, 13(4), e0195151-e0195151 (2018-04-13)
The sodium-independent cystine-glutamate antiporter plays an important role in extracellular cystine uptake. It comprises the transmembrane protein, xCT and its chaperone, CD98. Because glutathione is only weakly cell membrane permeable, cellular uptake of its precursor, cystine, is known to be
Yu Li et al.
Oncology letters, 19(1), 323-333 (2020-01-04)
Non-small cell lung cancer (NSCLC) has long been one of the most lethal types of cancer due to its lack of typical clinical symptoms at early stages and high risk of tumour recurrence, even following complete surgical resection. Multicourse chemotherapy
Chun Ge et al.
Scientific reports, 7(1), 3791-3791 (2017-06-21)
Adriamycin (ADR) induces the over-expression of P-glycoprotein (P-gp) and multiple drug resistance in breast cancer cells. However, the biochemical process and underlying mechanisms are not clear. Our previous study revealed that ADR increased reactive oxygen species (ROS) generation and decreased
Chaoli Huang et al.
Oncology reports, 40(4), 2363-2370 (2018-08-02)
The tumour‑suppressor protein p53 is a key regulator of multiple cellular processes and exerts its tumour‑suppressor function by inducing apoptotic cell death. However, emerging evidence indicates that p53 is also involved in inducing ferroptosis, which is a unique iron‑dependent form

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service