Skip to Content
MilliporeSigma

Skip To

D7069

DAPK1 (1-363), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DKFZp7811035

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 μg
Check Cart for Availability
$432.00

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200
Specific activity:
42-57 nmol/min·mg
Assay:
≥70% (SDS-PAGE)
Recombinant:
expressed in baculovirus infected Sf9 cells

$432.00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in baculovirus infected Sf9 cells

Quality Segment

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

42-57 nmol/min·mg

mol wt

~71 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... DAPK1(1612)

Biochem/physiol Actions

Death-associated protein kinase 1 (DAPK1) is a positive mediator of apoptosis induced by g-interferon. Activation of DAPK occurs via dephosphorylation of Ser-308 and subsequent association of calcium/calmodulin. DAPK is rapidly dephosphorylated in response to tumor necrosis factor or ceramide and then subsequently degraded via proteasome activity. The decline in DAPK expression is paralleled with increased caspase activity and cell apoptosis. Studies suggest that the apoptosis regulatory activities mediated by DAPK are controlled both by phosphorylation status and protein stability.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D7444S6948SRP5018
Gene Information

human ... DAPK1(1612)

Gene Information

human ... MAPK13(5603)

Gene Information

human ... SGK2(10110)

Gene Information

human ... DAPK2(23604)

assay

≥70% (SDS-PAGE)

assay

≥70% (SDS-PAGE)

assay

≥70% (SDS-PAGE)

assay

≥70% (SDS-PAGE)

specific activity

42-57 nmol/min·mg

specific activity

220-298 nmol/min·mg

specific activity

70-94 nmol/min·mg

specific activity

115-156 nmol/min·mg

recombinant

expressed in baculovirus infected Sf9 cells

recombinant

expressed in baculovirus infected Sf9 cells

recombinant

expressed in baculovirus infected Sf9 cells

recombinant

expressed in baculovirus infected Sf9 cells

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters