Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS480136

Corning® LSE rotors for LSE compact centrifuges

fixed angle rotor for 6 x 50 mL tubes, 6,000 rpm, 1/cs

Synonym(s):

fixed angle centrifuge rotor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103909
NACRES:
NB.15

feature

to hold 6 x 50 mL tubes

packaging

case of 1
pack of 1

manufacturer/tradename

Corning 480136

parameter

200-6000 max. RCF

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models LSE Compact

Looking for similar products? Visit Product Comparison Guide

Legal Information

Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Protocols

HPLC separation of 17 important cannabinoids including CBD, delta 9 THC and THCA. Read the application note.

HPLC separation of 17 important cannabinoids including CBD, delta 9 THC and THCA. Read the application note.

HPLC separation of 17 important cannabinoids including CBD, delta 9 THC and THCA. Read the application note.

HPLC separation of 17 important cannabinoids including CBD, delta 9 THC and THCA. Read the application note.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service