Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS4581

Corning® High Content Imaging Microplate

flat bottom, sterile, glass bottom, lid, pack of 10

Synonym(s):

384 multiwell plate, 384 well microplate, 384 well microtiter plate, 384 well plate, imaging plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13

material

black wells (with 0.2 mm glass bottom)
clear bottom
flat bottom
flat bottom wells

sterility

sterile

feature

lid
skirt
glass bottom

packaging

pack of 10
pack of 20 plates

manufacturer/tradename

Corning 4581

size

384 wells

well volume

82 μL

Looking for similar products? Visit Product Comparison Guide

General description

  • Corning® 384-well high content image screening microplates are best suited for performing high content cell-based assays using imaging systems.
  • The 0.2 mm thick glass bottom provides an optically clear surface, thus minimizing autofocus time and increasing throughput.
  • The microplate well geometry reduces reagent loss due to wicking and prevents cells from clumping in the corners.
  • Black color plates offer low background fluorescence and minimal crosstalk that provides the highest possible optical quality for cell-based assays.

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

A frequently asked questions guide for users of the Corning® high content screening microplates.

A frequently asked questions guide for users of the Corning® high content screening microplates.

A frequently asked questions guide for users of the Corning® high content screening microplates.

A frequently asked questions guide for users of the Corning® high content screening microplates.

Protocols

ICC Cell Capture Imaging Reagent simplifies imaging, ideal for rare cell and low cell count samples.

ICC Cell Capture Imaging Reagent simplifies imaging, ideal for rare cell and low cell count samples.

ICC Cell Capture Imaging Reagent simplifies imaging, ideal for rare cell and low cell count samples.

ICC Cell Capture Imaging Reagent simplifies imaging, ideal for rare cell and low cell count samples.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service