Key Documents
CLS3200
Corning® CellCube® Module 10-Layer with 8500 cm2 growth surface
polystyrene (growth surface), sterile, case of 2
About This Item
UNSPSC Code:
41122104
NACRES:
NB.13
material
polystyrene (growth surface)
sterility
sterile
feature
graduations: no
packaging
pack of 1
case of 2
manufacturer/tradename
Corning 3200
surface area
8500 cm2
working volume
600 mL
General description
The CellCube Module is an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment. It is comprised of a series of parallel, styrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates.
Each culture plate receives proprietary tissue culture treatment prior to assembly to ensure dense, uniform cell growth.
These CellCube Modules provide:
  • a series of parallel, polystyrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates.
  • 8,500cm2 polystyrene growth surface optimally treated for the growth of attachment dependent cells.
  • a perfused environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen.
  • an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment
Legal Information
CELLCUBE is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.