Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS3005

Corning® single use spatulas

Round end/spoon

Synonym(s):

Disposable Spatulas

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122403
NACRES:
NB.14

sterility

sterile

manufacturer/tradename

Corning 3005

L

248 mm

Looking for similar products? Visit Product Comparison Guide

General description

Corning single use spatulas have been designed to save time and provide contamination-free samples. Eliminates need to recycle and sterilize reusable spatulas.

Each spatula is individually packaged, RNase/DNase free, non-pyrogenic and antistatic.

Features and Benefits

  • Corning® single use spatulas are made of Polystyrene.
  • Color concentrate blend with 248 mm length.
  • Spatulas have a round end flat blade at one end and a spoon at the other end.
  • Visually Inspected for accurate labeling, product configuration, seal, and barrier integrity.

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service