Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COR-L24

96020717, human lymph node, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

COR-L24, 96020717

biological source

human lymph node

growth mode

Aggregates in suspension

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human lung small cell carcinoma

Cell Line Description

The cell line COR-L24 was established from a lymph node biopsy of an untreated 71 year old male and was diagnosed as small cell lung carcinoma (SCLC). Bone marrow taken from the same patient contained hypercellular marrow consistent with normal haemopoietic cells and no evidence of neoplasm. In culture, this xenograft forming cell line grows as floating aggregates of packed cells with irregular shape.

Application

Cytotoxicity studies, study of small cell lung cancer

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11
D13S317: 12
D16S539: 12
D5S818: 11
D7S820: 11
THO1: 7,9.3
TPOX: 8
vWA: 16,17

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9 x 100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service