Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G-292 Clone A141B1

90110522, human bone, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

G-292 Clone A141B1, 90110522

biological source

human bone

growth mode

Adherent

karyotype

Modal no. 57

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian osteosarcoma

Cell Line Description

Derived from a primary osteosarcoma of a 9 year old Caucasian female. Cells exhibit the type B phenotype for G6PD and the karyotype is subtetraploid with chromosome markers.

Application

Tumourigenicity studies

Culture Medium

McCoy′s 5a + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:5 i.e. seeding at 2-5x10,000 cells/cm2 using 0.25% trypsin/0.02% EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service