Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

K6H6/B5

89101606

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Human x Mouse Hybridoma hybrid

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse x Human hybrid myeloma

Cell Line Description

Derived by a fusion of NS-1 - Ag4 myeloma cells with cells from a human nodular lymphoma. The cells are HPRT- and do not secrete immunoglobulin. They are used as a fusion partner for human B lymphocytes.

Application

Fusion partner and hybridisation studies

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 1 x 105 to 1 x 106 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Selection of the right fusion partner cell line is critical to obtain desired attributes of the hybridoma. Explore hybridoma fusion partners cell lines.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service