Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SNL 76/7 Cell Line from mouse

7032801, mouse esophagus, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

SNL 76/7 Cell Line from mouse, 07032801

biological source

mouse esophagus

description

Mouse SIM strain embryonic fibroblast

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse SIM strain embryonic fibroblast

Cell Line Description

SNL 76/7 clonally-derived from a STO cell line that expresses both G418 resistance and leukaemic inhibitory factor (LIF). The STO cell line was transfected with a G418-resistance cassette, RV4.0 (Thomas and Capecchi 1987), and a LIF expression construct (Williams et al., 1988). This cell line can be used as a feeder layer to support the growth of mouse embryonic stem cells. Inclusion of G418 in the media is not necessary for normal cell growth.

Culture Medium

Knockout D-MEM (Invitrogen) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Split confluent cultures 1:5-1:15 i.e. seeding at 2-4x10,000 cells/cm2 using trypsin/EDTA;5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service