Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CHP-134

6122002, human nerve (neuroblastoma), Neuronal

Synonym(s):

CHP134 Cells, Children′s Hospital of Philadelphia-134 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

CHP-134, 06122002

biological source

human nerve (neuroblastoma)

growth mode

Adherent

karyotype

Not specified

morphology

Neuronal

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human neuroblastoma tumour mass of left adrenal gland

Cell Line Description

A neuroblastoma cell line established from a tumour of the left adrenal area of an 18-month-old male patient. The patient was previously treated with vincristine, cyclophosphamide, dimethyltriazenoimidazole carboxamide, adriamycin, and irradiation. The cells grow as adherent colonies consisting of neuroblast-like cells forming dense mounds. Individual cells radiate from the mounds which reach confluency after routine passaging for several months. This cell line has been reported to have tyrosine hydroxylase activity.

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 9,13
D13S317: 8,11
D16S539: 11,13
D5S818: 11,12
D7S820: 9
THO1: 9,9.3
TPOX: 8
vWA: 18,19

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split confluent cultures 1:2 to 1:10 every 2-3 days, use gentle agitation to dislodge cells, trypsin/EDTA should not be necessary as cells only adhere loosely; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service