Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C-40020

Sigma-Aldrich

HEPES Buffered Saline Solution

Hepes BSS (30 mM), 250 ml

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

form

aqueous solution

packaging

pkg of 250 ml

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

General description

HEPES Buffered Saline Solution

Application

HEPES Buffered Saline Solution is prepared from raw materials of the highest quality using tissue culture grade water and provided in a ready-to-use format. It is a component in the PromoCell DetachKits designed for the safe and efficient detachment of primary human cells in routine subculturing.

Quality

All lots of PromoCell HEPES Buffered Saline Solution are subjected to comprehensive quality control tests. Each lot is routinely tested for absence of cytotoxicity, and physical parameters such as osmolality and pH level. Approved in-house lots are used as a reference. In addition, all lots have been tested for the absence of microbial contaminants (fungi, bacteria).

Storage and Stability

Store HEPES Buffered Saline Solution at -20°C immediately after arrival. If stored properly, the product is stable until the expiry date stated on the label.

Subculture Routine

Click here for more information.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service