Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C-28030

Sigma-Aldrich

Mononuclear Cell Medium

Ready-to-use kit including Basal Medium and SupplementMix, 500 ml

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.81

form

liquid

packaging

pkg of 500 ml

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

General description

Mononuclear Cell Medium (Ready-to-use)

Application

Mononuclear Cell Medium is a very rich short-term maintenance medium, which provides utmost cell viability for Mononuclear Cells from umbilical cord blood, adult peripheral blood and other sources, e.g. bone marrow. It is suitable to maintain Mononuclear Cells for up to 48 hours before proceeding with your experiments.Recommended for:Human Mononuclear Cells from Cord Blood (hMNC-CB)Human Mononuclear Cells from Peripheral Blood (hMNC-PB)

Quality

All lots of PromoCell Mononuclear Cell Medium are subjected to comprehensive quality control tests using primary human mononuclear cells from cord blood and adult peripheral blood. Each lot of PromoCell Mononuclear Cell Medium is checked for unaltered shortterm maintenance of highly viable mononuclear cells. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Preparation Note

The Medium consists of a bottle of Basal Medium and one vial of SupplementMix. Adding the SupplementMix to the Basal Medium results in the complete Medium.

Storage and Stability

Store the Basal Medium at 4 to 8°C in the dark and the SupplementMix at -20°C immediately after arrival. Do not freeze the Basal Medium. If stored properly, the products are stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C. Note: The SupplementMix is delivered thawed and can be frozen after arrival without losing any activity

Subculture Routine

Click here for more information.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Umbilical cord blood and adult peripheral blood mononuclear cells with optimized maintenance medium for short-term cultivation.

Umbilical cord blood and adult peripheral blood mononuclear cells with optimized maintenance medium for short-term cultivation.

Umbilical cord blood and adult peripheral blood mononuclear cells with optimized maintenance medium for short-term cultivation.

Umbilical cord blood and adult peripheral blood mononuclear cells with optimized maintenance medium for short-term cultivation.

See All

Protocols

Generate mature human dendritic immune cells from PBMCs or CD34+ stem cell derived monocytes in 7 days in defined and serum-free dendritic cell culture media. Explore over 350 PromoCell products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service