Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

B354-05

Sigma-Aldrich

Bovine Aortic Smooth Muscle Cells: BAOSMC (Cryovial)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

bovine artery (USDA-inspected cattle)

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

karyotype

2n = 60

morphology

smooth muscle

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cardiovascular diseases

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Bovine large artery smooth muscle cells (BAOSMC) provide an excellent model system to study all aspects of cardiovascular function and disease, especially those related to mechanisms of hyperplasia and hypertrophy of intimal smooth muscle cells leading to vascular occlusion in atherosclerosis and stent restenosis.

BAOSMC have been utilized in a number of research studies, for example, to:
  • Elucidate RhoA-dependent serum response signaling pathways (Gudi, 2002); the role of FAK in thrombospondin-1 induced migration of vascular smooth muscle cells (Wang, 2008)
  • Demonstrate the relationship between the disturbed flow and proliferation of smooth muscle cells, leading to intima hyperplasia (Fan, 2010); and study interactions between lipoproteins and extracellular matrix, as well as mechanisms of lipoprotein aggregation contributing to atherosclerosis (Sakr, 2001)
  • Construct an expression cassette to maximize targeted transgene expression (Dronadula, 2010)
  • Test new technologies, such as on-chip contact imaging technique (Liu, 2004; Ji, 2005, 2007; Prakash, 2007), handheld fluorometers (Nelson, 2009) and a “nose on a chip” olfactory sensor (Patel, 2011)
  • Study the behavior of smooth muscle cells in 3d environment (Du, 2008) and smooth muscle/endothelial co-cultures (Niwa, 2007; Fan, 2008; Mathura, 2013); describe an origami-like method of generating three-dimensional (3D) cell-laden microstructures (Kuribayashi-Shigetomi, 2012); and design materials for tissue engineering and cardiovascular implants (Mathews, 2006, 2008)

    Characterization: positive for smooth muscle cell specific alpha-actin expression.

Cell Line Origin

Artery

Application

aortic wall contraction, role of smc under normal conditions, blood pressure, distribution of oxygenated blood through systemic circulation

Components

Bovine Smooth Muscle Cell Basal Medium containing 10% FBS & 10% DMSO

Preparation Note

  • 2nd passage, >500,000 cells in Bovine Smooth Muscle Cell Basal Medium containing 10% FBS & 10% DMSO
  • Can be cultured at least 16 doublings

Subculture Routine

Please refer to the BAOSMC Culture Protocol.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service