Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AXYSCT5MLS

Screw-cap tubes and caps, self-standing, unassembled

capacity 5 mL, blue, sterile

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.11

material

blue
conical bottom tube
screw top cap

sterility

sterile

feature

graduations

packaging

pack of 25 ea
case of 500 ea

manufacturer/tradename

Axygen Inc SCT-5ML-S
Corning

capacity

5 mL

color

clear

fitting

cap (screw cap)

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Unassembled and made up of virgin polypropylene
  • Free from RNase-/DNase and human DNA
  • Large, frosted writing area with graduation at intervals of 1.0 mL
  • Tight fit between the tube and the cap ensures a leak-proof seal
  • Sterile and nonpyrogenic in nature

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service